Product Details

SNP ID
rs7771595
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:84136078 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TGTCTTAAGCAGCCATTCCTGCTTT[A/C]TTTGCTGACAAGGCCTTGATATTGT
Phenotype
MIM: 610201 MIM: 615410
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
CEP162 PubMed Links

Gene Details

Gene
CEP162
Gene Name
centrosomal protein 162
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001286206.1 Intron NP_001273135.1
NM_014895.3 Intron NP_055710.2
XM_005248674.3 Intron XP_005248731.1
XM_005248678.3 Intron XP_005248735.1
XM_006715380.2 Intron XP_006715443.1
XM_011535592.2 Intron XP_011533894.1
XM_011535594.2 Intron XP_011533896.1
XM_017010483.1 Intron XP_016865972.1
XM_017010484.1 Intron XP_016865973.1
Gene
MRAP2
Gene Name
melanocortin 2 receptor accessory protein 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_138409.2 Intron NP_612418.2
XM_011535400.2 Intron XP_011533702.1
XM_017010219.1 Intron XP_016865708.1
XM_017010220.1 Intron XP_016865709.1
XM_017010221.1 Intron XP_016865710.1

View Full Product Details