Product Details

SNP ID
rs5746129
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:159684911 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CAAGCAGCAATTTGTAAGTGTCCCC[A/G]TTCCTTATTGAAACCAAGCCAACCC
Phenotype
MIM: 147460
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
SOD2 PubMed Links

Gene Details

Gene
SOD2
Gene Name
superoxide dismutase 2, mitochondrial
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000636.3 2156 Missense Mutation CGG,TGG R156W NP_000627.2
NM_001024465.2 2156 Missense Mutation CGG,TGG R156W NP_001019636.1
NM_001024466.2 2156 Missense Mutation CGG,TGG R117W NP_001019637.1
NM_001322814.1 2156 Missense Mutation CGG,TGG R117W NP_001309743.1
NM_001322815.1 2156 Intron NP_001309744.1
NM_001322816.1 2156 Intron NP_001309745.1
NM_001322817.1 2156 Missense Mutation CGG,TGG R110W NP_001309746.1
NM_001322819.1 2156 Missense Mutation CGG,TGG R110W NP_001309748.1
NM_001322820.1 2156 Missense Mutation CGG,TGG R110W NP_001309749.1

View Full Product Details