Product Details

SNP ID
rs6933765
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:10399118 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGCAGCCATCAGCCTTCCTATTCAA[A/C]ATGTCATAAAGACCCTCTTCCCTGC
Phenotype
MIM: 107580
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
TFAP2A PubMed Links

Gene Details

Gene
TFAP2A
Gene Name
transcription factor AP-2 alpha
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001032280.2 Intron NP_001027451.1
NM_001042425.1 Intron NP_001035890.1
NM_003220.2 Intron NP_003211.1
XM_006715175.2 Intron XP_006715238.1
XM_011514833.2 Intron XP_011513135.1
XM_017011232.1 Intron XP_016866721.1

View Full Product Details