Product Details

SNP ID
rs7804972
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:64679085 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ATGGTGTAGTCTGGATTGAAGAGAC[A/G]ACCCAAGCAGGCTTTGTGTGAGCAG
Phenotype
MIM: 603989
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
MIR6839 PubMed Links

Gene Details

Gene
MIR6839
Gene Name
microRNA 6839
There are no transcripts associated with this gene.

Gene
ZNF107
Gene Name
zinc finger protein 107
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001013746.2 Intron NP_001013768.1
NM_001282359.1 Intron NP_001269288.1
NM_001282360.1 Intron NP_001269289.1
NM_016220.4 Intron NP_057304.1
XM_017012283.1 Intron XP_016867772.1
XM_017012284.1 Intron XP_016867773.1
XM_017012285.1 Intron XP_016867774.1
XM_017012286.1 Intron XP_016867775.1

View Full Product Details