Product Details
- SNP ID
-
rs11555922
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.7:16753998 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TTGCGGGGGCTTCGCGTGTTCCAAG[A/G]ACTGCCTGTGCGCCCTCAACCTGCT
- Phenotype
-
MIM: 613139
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
TSPAN13
PubMed Links
Gene Details
- Gene
- TSPAN13
- Gene Name
- tetraspanin 13
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_014399.3 |
273 |
Missense Mutation |
AAC,GAC |
N11D |
NP_055214.1 |
View Full Product Details