Product Details

SNP ID
rs11551168
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:2355226 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGGTCGCATGCTGAGCCCCCTGTCC[C/T]GGCACAGGGCGAGGCCCCAGGAGAG
Phenotype
MIM: 603917 MIM: 614905
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
EIF3B PubMed Links

Gene Details

Gene
EIF3B
Gene Name
eukaryotic translation initiation factor 3 subunit B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001037283.1 687 Missense Mutation CCG,CTG P102L NP_001032360.1
NM_003751.3 687 Missense Mutation CCG,CTG P102L NP_003742.2
XM_011515599.1 687 Missense Mutation CCG,CTG P102L XP_011513901.1
XM_011515600.1 687 Missense Mutation CCG,CTG P102L XP_011513902.1
XM_011515601.1 687 Intron XP_011513903.1
XM_017012752.1 687 Missense Mutation CCG,CTG P102L XP_016868241.1
XM_017012753.1 687 Missense Mutation CCG,CTG P102L XP_016868242.1
Gene
SNX8
Gene Name
sorting nexin 8
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_013321.3 687 Intron NP_037453.1
XM_011515329.2 687 Intron XP_011513631.1
XM_011515330.1 687 Intron XP_011513632.1
XM_017012083.1 687 Intron XP_016867572.1
XM_017012084.1 687 Intron XP_016867573.1

View Full Product Details