Product Details
- SNP ID
-
rs10280681
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.7:32978850 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- GTAGTTATTTTTGAATACTTTATAC[C/T]TGAATACTGTGCTCGGCTATCCAAG
- Phenotype
-
MIM: 616257
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
FKBP9
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs150781767] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- FKBP9
- Gene Name
- FK506 binding protein 9
View Full Product Details