Product Details
- SNP ID
-
rs13228799
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.7:142740720 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- ATGATGTTCTGGTACCGTCAGCAAC[C/T]TGGACAGAGCCTGACACTGATCGCA
- Phenotype
-
MIM: 276000
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
PRSS1
PubMed Links
Gene Details
- Gene
- PRSS1
- Gene Name
- protease, serine 1
View Full Product Details