Product Details

SNP ID
rs2951313
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.7:144187029 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTGAGAGCATCTGATGGGCTCTAGC[A/G]AGGGAGACAGAGCTGCGGGGGACAG
Phenotype
MIM: 608910
Polymorphism
A/G, Transition Substitution
Allele Nomenclature
Literature Links
ARHGEF35 PubMed Links

Gene Details

Gene
ARHGEF35
Gene Name
Rho guanine nucleotide exchange factor 35
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001003702.2 Intron NP_001003702.2
Gene
CTAGE4
Gene Name
CTAGE family member 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_198495.2 Intron NP_940897.2
Gene
LOC101928605
Gene Name
uncharacterized LOC101928605
There are no transcripts associated with this gene.

View Full Product Details