Product Details
- SNP ID
-
rs7789439
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.7:101028875 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TGCACTCCAGCCTGGGCGATACAGA[C/G]AGACACTGTCTCTTAAAAATTAATA
- Phenotype
-
MIM: 604609
MIM: 608424
- Polymorphism
- C/G, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
MUC12
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs75024434] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- MUC12
- Gene Name
- mucin 12, cell surface associated
There are no transcripts associated with this gene.
- Gene
- MUC17
- Gene Name
- mucin 17, cell surface associated
View Full Product Details