Product Details

SNP ID
hCV43154128
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:76048176 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CGCTCCAGCCATGCTCTCCGCCCTC[G/T]CCCGGCCTGCCAGCGCTGCTCTCCG
Phenotype
MIM: 154100 MIM: 616695
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
MDH2 PubMed Links

Gene Details

Gene
MDH2
Gene Name
malate dehydrogenase 2
There are no transcripts associated with this gene.

Gene
STYXL1
Gene Name
serine/threonine/tyrosine interacting-like 1
There are no transcripts associated with this gene.

View Full Product Details