Product Details

SNP ID
rs7845663
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:42753420 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TTTTTGAAGAAGATTTCTCCGGCAC[A/G]TTTTAAAGCAAATATCAGCTGGGTG
Phenotype
MIM: 606888
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CHRNA6 PubMed Links

Gene Details

Gene
CHRNA6
Gene Name
cholinergic receptor nicotinic alpha 6 subunit
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001199279.1 Intron NP_001186208.1
NM_004198.3 Intron NP_004189.1

View Full Product Details