Product Details

SNP ID
rs7018049
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:95139210 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTGGATTTACAGGAGCCCTTTGCAT[A/G]TTACAAATGTTGGCCGGGTGCGGTG
Phenotype
MIM: 615208
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
PLEKHF2 PubMed Links

Gene Details

Gene
PLEKHF2
Gene Name
pleckstrin homology and FYVE domain containing 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_024613.3 Intron NP_078889.1

View Full Product Details