Product Details

SNP ID
rs11542565
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:128689619 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGCGGCCAAAGTCAGTAAAAAGGAG[C/T]TCAACTCCAACCACGACGGGGCCGA
Phenotype
MIM: 600960
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
SET PubMed Links

Gene Details

Gene
SET
Gene Name
SET nuclear proto-oncogene
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001122821.1 433 Intron NP_001116293.1
NM_001248000.1 433 Intron NP_001234929.1
NM_001248001.1 433 Intron NP_001234930.1
NM_003011.3 433 Missense Mutation CTC,TTC L13F NP_003002.2
XM_017015013.1 433 Intron XP_016870502.1
XM_017015014.1 433 Intron XP_016870503.1
XM_017015015.1 433 Intron XP_016870504.1
XM_017015016.1 433 Missense Mutation CTC,TTC L13F XP_016870505.1

View Full Product Details