Product Details

SNP ID
rs11549849
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:78035522 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATATCTCCTTTTAGGTCATTGATGT[C/T]CTTCACCCCGGGAAGGCGACAGTGC
Phenotype
MIM: 614258 MIM: 602412
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
POLR3A PubMed Links

Gene Details

Gene
POLR3A
Gene Name
RNA polymerase III subunit A
There are no transcripts associated with this gene.

Gene
RPS24
Gene Name
ribosomal protein S24
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001026.4 223 Intron NP_001017.1
NM_001142282.1 223 Intron NP_001135754.1
NM_001142283.1 223 Intron NP_001135755.1
NM_001142284.1 223 Intron NP_001135756.1
NM_001142285.1 223 Silent Mutation GTC,GTT V27V NP_001135757.1
NM_033022.3 223 Intron NP_148982.1

View Full Product Details