Product Details
- SNP ID
-
hCV43386390
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.11:95067756 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CGCTACAGCCGCTCTCCCTACAGCA[G/T]ATCTCGCTACAGGGAATCTCGCTAC
- Phenotype
-
MIM: 603269
- Polymorphism
- G/T, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
SRSF8
PubMed Links
Gene Details
- Gene
- SRSF8
- Gene Name
- serine and arginine rich splicing factor 8
There are no transcripts associated with this gene.
View Full Product Details