Product Details

SNP ID
rs7941001
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:32587977 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCTATATGTATACACAAAGTTTTCC[G/T]TAATTAAGAAAGTATCCTAGTCCTA
Phenotype
MIM: 609641
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
EIF3M PubMed Links

Gene Details

Gene
EIF3M
Gene Name
eukaryotic translation initiation factor 3 subunit M
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001307929.1 Intron NP_001294858.1
NM_006360.5 Intron NP_006351.2
XM_011519840.2 Intron XP_011518142.1

View Full Product Details