Product Details
- SNP ID
-
rs7113992
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.11:4571522 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- CCTGGAGCAGTGCTAGGATGGAATA[C/T]TCTGATGGAAGTGTTTTGTAATCAG
- Phenotype
-
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
C11orf40
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs141600462] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- C11orf40
- Gene Name
- chromosome 11 open reading frame 40
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_144663.1 |
555 |
Silent Mutation |
GAA,GAG |
E185E |
NP_653264.1 |
View Full Product Details