Product Details
- SNP ID
-
rs11545529
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.11:47259862 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CCAGGCCCAGGGAGGCAGCAGCTGC[A/G]TCCTCAGAGAGGAAGCCAGGATGCC
- Phenotype
-
MIM: 603584
MIM: 602423
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
MADD
PubMed Links
Gene Details
- Gene
- MADD
- Gene Name
- MAP kinase activating death domain
There are no transcripts associated with this gene.
- Gene
- NR1H3
- Gene Name
- nuclear receptor subfamily 1 group H member 3
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001130101.2 |
177 |
Missense Mutation |
ATC,GTC |
I39V |
NP_001123573.1 |
NM_001130102.2 |
177 |
UTR 5 |
|
|
NP_001123574.1 |
NM_001251934.1 |
177 |
Missense Mutation |
ATC,GTC |
I45V |
NP_001238863.1 |
NM_001251935.1 |
177 |
Missense Mutation |
ATC,GTC |
I45V |
NP_001238864.1 |
NM_005693.3 |
177 |
Missense Mutation |
ATC,GTC |
I39V |
NP_005684.2 |
XM_005252705.1 |
177 |
Missense Mutation |
ATC,GTC |
I39V |
XP_005252762.1 |
XM_005252706.1 |
177 |
Missense Mutation |
ATC,GTC |
I39V |
XP_005252763.1 |
XM_005252707.4 |
177 |
Missense Mutation |
ATC,GTC |
I39V |
XP_005252764.1 |
XM_005252709.1 |
177 |
UTR 5 |
|
|
XP_005252766.1 |
XM_005252710.1 |
177 |
UTR 5 |
|
|
XP_005252767.1 |
XM_005252713.3 |
177 |
Missense Mutation |
ATC,GTC |
I39V |
XP_005252770.1 |
XM_005252715.2 |
177 |
Missense Mutation |
ATC,GTC |
I39V |
XP_005252772.1 |
XM_005252716.3 |
177 |
UTR 5 |
|
|
XP_005252773.1 |
XM_005252718.3 |
177 |
UTR 5 |
|
|
XP_005252775.1 |
XM_006718112.1 |
177 |
Missense Mutation |
ATC,GTC |
I39V |
XP_006718175.1 |
XM_006718113.1 |
177 |
Missense Mutation |
ATC,GTC |
I39V |
XP_006718176.1 |
XM_006718115.1 |
177 |
UTR 5 |
|
|
XP_006718178.1 |
XM_006718116.1 |
177 |
UTR 5 |
|
|
XP_006718179.1 |
XM_011519805.2 |
177 |
Missense Mutation |
ATC,GTC |
I39V |
XP_011518107.1 |
XM_011519806.1 |
177 |
UTR 5 |
|
|
XP_011518108.1 |
XM_011519807.1 |
177 |
UTR 5 |
|
|
XP_011518109.1 |
XM_011519808.2 |
177 |
Intron |
|
|
XP_011518110.1 |
XM_017017056.1 |
177 |
UTR 5 |
|
|
XP_016872545.1 |
XM_017017057.1 |
177 |
UTR 5 |
|
|
XP_016872546.1 |
XM_017017058.1 |
177 |
UTR 5 |
|
|
XP_016872547.1 |
View Full Product Details