Product Details
- SNP ID
-
rs7305183
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.12:27782847 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- AAATTACACACAAAACTGTGGAGTA[A/C]AAAGAGTAAATGTCTGCCCTCAGGG
- Phenotype
-
- Polymorphism
- A/C, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
KLHL42
PubMed Links
Gene Details
- Gene
- KLHL42
- Gene Name
- kelch like family member 42
View Full Product Details