Product Details

SNP ID
rs7974701
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:28195579 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TTTGGCTAAAACCTCAGTCCTTATT[A/G]TGGTAGCGTTATAGTGTATCTTGAA
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CCDC91 PubMed Links

Gene Details

Gene
CCDC91
Gene Name
coiled-coil domain containing 91
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
XM_005253413.1 Intron XP_005253470.1
XM_005253414.1 Intron XP_005253471.1
XM_005253415.1 Intron XP_005253472.1
XM_006719103.3 Intron XP_006719166.1
XM_006719104.2 Intron XP_006719167.1
XM_006719105.1 Intron XP_006719168.1
XM_011520729.2 Intron XP_011519031.1
XM_011520731.2 Intron XP_011519033.1
XM_011520733.2 Intron XP_011519035.1
XM_011520734.1 Intron XP_011519036.1
XM_011520735.1 Intron XP_011519037.1
XM_017019569.1 Intron XP_016875058.1
XM_017019570.1 Intron XP_016875059.1
XM_017019571.1 Intron XP_016875060.1
XM_017019572.1 Intron XP_016875061.1
XM_017019573.1 Intron XP_016875062.1
XM_017019574.1 Intron XP_016875063.1
XM_017019575.1 Intron XP_016875064.1
XM_017019576.1 Intron XP_016875065.1
XM_017019577.1 Intron XP_016875066.1
Gene
LOC729291
Gene Name
translation initiation factor IF-2
There are no transcripts associated with this gene.

View Full Product Details