Product Details

SNP ID
rs11608362
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:10433481 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATTTCATGATCTGACTTGTATCACA[C/T]GCATTCATTCACTTTGTGATCAATC
Phenotype
MIM: 161555 MIM: 602891
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
KLRC1 PubMed Links

Gene Details

Gene
KLRC1
Gene Name
killer cell lectin like receptor C1
There are no transcripts associated with this gene.

Gene
KLRC2
Gene Name
killer cell lectin like receptor C2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002260.3 Intron NP_002251.2

View Full Product Details