Product Details

SNP ID
rs11548379
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:120198605 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCTCTGTGATATCAAGCACTTCAGG[G/T]TTGTAGATGCTGCCATTGTCGAACA
Phenotype
MIM: 605614 MIM: 180510
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
GCN1 PubMed Links

Gene Details

Gene
GCN1
Gene Name
GCN1, eIF2 alpha kinase activator homolog
There are no transcripts associated with this gene.

Gene
PXN-AS1
Gene Name
PXN antisense RNA 1
There are no transcripts associated with this gene.

Gene
RPLP0
Gene Name
ribosomal protein lateral stalk subunit P0
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001002.3 Intron NP_000993.1
NM_053275.3 Intron NP_444505.1

View Full Product Details