Product Details

SNP ID
rs6489886
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:112980202 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ATGGATACAATGTGAAATGATCAGG[A/G]TAGTTAGGATATCCATCACCTCAAA
Phenotype
MIM: 603350 MIM: 603351
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
OAS2 PubMed Links

Gene Details

Gene
OAS2
Gene Name
2'-5'-oligoadenylate synthetase 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001032731.1 Intron NP_001027903.1
NM_002535.2 Intron NP_002526.2
NM_016817.2 Intron NP_058197.2
Gene
OAS3
Gene Name
2'-5'-oligoadenylate synthetase 3
There are no transcripts associated with this gene.

View Full Product Details