Product Details

SNP ID
rs7299040
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:118380660 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AGAAAGAATCTTGAGGGCAATGGAC[A/C]CAGGGGAGGGAATGAGGCCGTACAA
Phenotype
MIM: 608250 MIM: 616711
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
SUDS3 PubMed Links

Gene Details

Gene
SUDS3
Gene Name
SDS3 homolog, SIN3A corepressor complex component
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_022491.2 Intron NP_071936.2
XM_011538671.2 Intron XP_011536973.1
XM_017019817.1 Intron XP_016875306.1
XM_017019818.1 Intron XP_016875307.1
XM_017019819.1 Intron XP_016875308.1
XM_017019820.1 Intron XP_016875309.1
Gene
TAOK3
Gene Name
TAO kinase 3
There are no transcripts associated with this gene.

View Full Product Details