Product Details

SNP ID
rs7323661
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.13:99507108 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TATGCCTTAGCTAAATTTCATAACA[C/T]CTCTGTGATATTTAGATGAGGAAAC
Phenotype
MIM: 604678
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
LINC00449 PubMed Links

Gene Details

Gene
LINC00449
Gene Name
long intergenic non-protein coding RNA 449
There are no transcripts associated with this gene.

Gene
LINC01232
Gene Name
long intergenic non-protein coding RNA 1232
There are no transcripts associated with this gene.

Gene
TM9SF2
Gene Name
transmembrane 9 superfamily member 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004800.2 Intron NP_004791.1

View Full Product Details