Product Details

SNP ID
rs7982446
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.13:109758368 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TTATTTTGACACTAACAATTAGCTA[A/G]ATGTGGCTGAATTATCTCATGTTAA
Phenotype
MIM: 600797
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
IRS2 PubMed Links

Gene Details

Gene
IRS2
Gene Name
insulin receptor substrate 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003749.2 Intron NP_003740.2

View Full Product Details