Product Details

SNP ID
rs11553032
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:44715449 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TACTCCAAAGATTCAGGTTTACTCA[C/T]GTCATCCAGCAGAGAATGGAAAGTC
Phenotype
MIM: 109700 MIM: 614661
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
B2M PubMed Links

Gene Details

Gene
B2M
Gene Name
beta-2-microglobulin
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004048.2 149 Missense Mutation CGT,TGT R32C NP_004039.1
XM_005254549.3 149 Missense Mutation CGT,TGT R32C XP_005254606.1
Gene
PATL2
Gene Name
PAT1 homolog 2
There are no transcripts associated with this gene.

View Full Product Details