Product Details

SNP ID
hCV43558966
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:73444252 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATGCAGTTTGTCACATGGATTGCCT[C/G]TAACTTTCAGGAAAGATTGCTCTGT
Phenotype
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
LOC107984728 PubMed Links

Gene Details

Gene
LOC107984728
Gene Name
serine-aspartate repeat-containing protein I-like
There are no transcripts associated with this gene.

Gene
REC114
Gene Name
REC114 meiotic recombination protein
There are no transcripts associated with this gene.

View Full Product Details