Product Details

SNP ID
rs11555267
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:67150309 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AGGCCGGCGGGGCTGCACCCGCGTC[G/T]GGTAACTGGAACGCGCGGGGTCCGG
Phenotype
MIM: 609077 MIM: 603500
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
B3GNT9 PubMed Links

Gene Details

Gene
B3GNT9
Gene Name
UDP-GlcNAc:betaGal beta-1,3-N-acetylglucosaminyltransferase 9
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_033309.2 498 Silent Mutation CCA,CCC P59P NP_171608.2
Gene
C16orf70
Gene Name
chromosome 16 open reading frame 70
There are no transcripts associated with this gene.

Gene
FBXL8
Gene Name
F-box and leucine rich repeat protein 8
There are no transcripts associated with this gene.

Gene
TRADD
Gene Name
TNFRSF1A associated via death domain
There are no transcripts associated with this gene.

View Full Product Details