Product Details

SNP ID
rs8062948
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:71645120 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AATACGACAATGATTGCACAAAACC[A/G]TAAGATATGAGCCCACTGTCTGGAT
Phenotype
MIM: 614094 MIM: 611066
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
MARVELD3 PubMed Links

Gene Details

Gene
MARVELD3
Gene Name
MARVEL domain containing 3
There are no transcripts associated with this gene.

Gene
PHLPP2
Gene Name
PH domain and leucine rich repeat protein phosphatase 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001289003.1 8121 UTR 3 NP_001275932.1
NM_015020.3 8121 UTR 3 NP_055835.2

View Full Product Details