Product Details

SNP ID
rs7224465
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:75018662 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAATAGAGAGATAAGACAAACTTGG[G/T]GGCTGAGAGACAAGAGTGAAAGACA
Phenotype
MIM: 603000
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
MRPL58 PubMed Links

Gene Details

Gene
MRPL58
Gene Name
mitochondrial ribosomal protein L58
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001303265.1 Intron NP_001290194.1
NM_001545.2 Intron NP_001536.1
XM_017024574.1 Intron XP_016880063.1

View Full Product Details