Product Details

SNP ID
rs11554352
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:15999963 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGGGCTGGCAGGAGGTCCGGGGCCC[A/G]AGGTGCAGGTGCCGCCATCCCGAGT
Phenotype
MIM: 613814 MIM: 614535
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
TTC19 PubMed Links

Gene Details

Gene
TTC19
Gene Name
tetratricopeptide repeat domain 19
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001271420.1 584 UTR 5 NP_001258349.1
NM_017775.3 584 Missense Mutation AAG,GAG K39E NP_060245.3
XM_017024801.1 584 Missense Mutation AAG,GAG K160E XP_016880290.1
XM_017024802.1 584 Missense Mutation AAG,GAG K160E XP_016880291.1
Gene
ZSWIM7
Gene Name
zinc finger SWIM-type containing 7
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001042697.1 584 Intron NP_001036162.1
NM_001042698.1 584 Intron NP_001036163.1

View Full Product Details