Product Details

SNP ID
rs11541057
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:4671825 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGGGTTGGGGGAGCAGAGGGCTCTT[C/T]TTCAACAAGCTCCTGGGGAGGGGGC
Phenotype
MIM: 609455
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
PELP1 PubMed Links

Gene Details

Gene
PELP1
Gene Name
proline, glutamate and leucine rich protein 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001278241.1 3392 Missense Mutation AAA,GAA K832E NP_001265170.1
NM_014389.2 3392 Missense Mutation AAA,GAA K1106E NP_055204.3

View Full Product Details