Product Details

SNP ID
hCV43721767
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:42659510 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CGCCCACGTCTGAAGAGCGATGCCC[A/C]GGGAGATCATCACCCTGCAGCTGGG
Phenotype
MIM: 605785
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
PLEKHH3 PubMed Links

Gene Details

Gene
PLEKHH3
Gene Name
pleckstrin homology, MyTH4 and FERM domain containing H3
There are no transcripts associated with this gene.

Gene
TUBG2
Gene Name
tubulin gamma 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001320509.1 263 Silent Mutation AGG,CGG R3R NP_001307438.1
NM_016437.2 263 Silent Mutation AGG,CGG R3R NP_057521.1

View Full Product Details