Product Details

SNP ID
rs2668707
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:64921216 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CAAAAAGGTAACCTTGCCCGCTCCC[C/T]ATACATTCCCACTAGGAGGTGTCAG
Phenotype
MIM: 616557
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
LRRC37A3 PubMed Links

Gene Details

Gene
LRRC37A3
Gene Name
leucine rich repeat containing 37 member A3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001303255.1 Intron NP_001290184.1
NM_199340.3 Intron NP_955372.2
XM_011524776.1 Intron XP_011523078.1
XM_011524777.1 Intron XP_011523079.1
XM_011524778.2 Intron XP_011523080.2
XM_017024593.1 Intron XP_016880082.1
XM_017024594.1 Intron XP_016880083.1
XM_017024595.1 Intron XP_016880084.1
XM_017024596.1 Intron XP_016880085.1
XM_017024597.1 Intron XP_016880086.1
XM_017024598.1 Intron XP_016880087.1
XM_017024599.1 Intron XP_016880088.1
XM_017024600.1 Intron XP_016880089.1
XM_017024601.1 Intron XP_016880090.1
XM_017024602.1 Intron XP_016880091.1
XM_017024603.1 Intron XP_016880092.1
XM_017024604.1 Intron XP_016880093.1
XM_017024605.1 Intron XP_016880094.1
XM_017024606.1 Intron XP_016880095.1
XM_017024607.1 Intron XP_016880096.1
XM_017024608.1 Intron XP_016880097.1

View Full Product Details