Product Details
- SNP ID
-
hCV43739491
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.17:67343047 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- ATTAAATTTCCTAATTTCAACTGGC[A/G]ATGGTGCTGAAGTTAAGACACAGAG
- Phenotype
-
MIM: 604450
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
PSMD12
PubMed Links
Gene Details
- Gene
- PSMD12
- Gene Name
- proteasome 26S subunit, non-ATPase 12
There are no transcripts associated with this gene.
View Full Product Details