Product Details
- SNP ID
-
rs11556546
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.17:44078550 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CAGCCTGGGCCTGCTCGGCCCCCAC[C/G]GACAGCAAGGCCATGGCGCTCACAG
- Phenotype
-
MIM: 611045
MIM: 605315
- Polymorphism
- C/G, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
G6PC3
PubMed Links
Gene Details
- Gene
- G6PC3
- Gene Name
- glucose-6-phosphatase catalytic subunit 3
There are no transcripts associated with this gene.
- Gene
- HDAC5
- Gene Name
- histone deacetylase 5
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001015053.1 |
3677 |
Silent Mutation |
TCC,TCG |
S1094S |
NP_001015053.1 |
NM_005474.4 |
3677 |
Silent Mutation |
TCC,TCG |
S1093S |
NP_005465.2 |
XM_005256905.1 |
3677 |
Silent Mutation |
TCC,TCG |
S1093S |
XP_005256962.1 |
XM_005256906.4 |
3677 |
Silent Mutation |
TCC,TCG |
S1093S |
XP_005256963.1 |
XM_011524149.1 |
3677 |
Silent Mutation |
TCC,TCG |
S1085S |
XP_011522451.1 |
XM_011524150.1 |
3677 |
Silent Mutation |
TCC,TCG |
S1084S |
XP_011522452.1 |
XM_017023988.1 |
3677 |
Silent Mutation |
TCC,TCG |
S1132S |
XP_016879477.1 |
XM_017023989.1 |
3677 |
Silent Mutation |
TCC,TCG |
S1132S |
XP_016879478.1 |
XM_017023990.1 |
3677 |
Silent Mutation |
TCC,TCG |
S1131S |
XP_016879479.1 |
XM_017023991.1 |
3677 |
Silent Mutation |
TCC,TCG |
S1131S |
XP_016879480.1 |
XM_017023992.1 |
3677 |
Silent Mutation |
TCC,TCG |
S1127S |
XP_016879481.1 |
XM_017023993.1 |
3677 |
Silent Mutation |
TCC,TCG |
S1123S |
XP_016879482.1 |
XM_017023994.1 |
3677 |
Silent Mutation |
TCC,TCG |
S1103S |
XP_016879483.1 |
XM_017023995.1 |
3677 |
Silent Mutation |
TCC,TCG |
S1084S |
XP_016879484.1 |
XM_017023996.1 |
3677 |
Silent Mutation |
TCC,TCG |
S1052S |
XP_016879485.1 |
View Full Product Details