Product Details

SNP ID
rs9284390
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.18:112535 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CAGCTCCCCCGGGGCCTCCGTTTCT[A/G]GGAAAGGGTTAGACACAGAGTAAGT
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
MIR8078 PubMed Links

Gene Details

Gene
MIR8078
Gene Name
microRNA 8078
There are no transcripts associated with this gene.

Gene
ROCK1P1
Gene Name
Rho associated coiled-coil containing protein kinase 1 pseudogene 1
There are no transcripts associated with this gene.

View Full Product Details