Product Details
- SNP ID
-
rs8096764
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.18:13732455 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- TACCTAGCTTTACATCCCTTTATTA[A/G]TGTAATTCCTTAAACAGTTTTCAGC
- Phenotype
-
MIM: 603514
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
FAM210A
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs75714597] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- FAM210A
- Gene Name
- family with sequence similarity 210 member A
There are no transcripts associated with this gene.
- Gene
- RNMT
- Gene Name
- RNA guanine-7 methyltransferase
View Full Product Details