Product Details

SNP ID
rs7240441
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.18:45986832 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ATTTAAGGACTTATGTTTAAATATA[A/G]TAGAAACCAGCTGATTCTTTCCTTT
Phenotype
MIM: 616046
Polymorphism
A/G, Transition Substitution
Allele Nomenclature
Literature Links
PSTPIP2 PubMed Links

Gene Details

Gene
PSTPIP2
Gene Name
proline-serine-threonine phosphatase interacting protein 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_024430.3 Intron NP_077748.3
XM_011526253.2 Intron XP_011524555.1
XM_011526254.1 Intron XP_011524556.1
XM_011526256.1 Intron XP_011524558.1

View Full Product Details