Product Details

SNP ID
rs12984442
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:46635517 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACAAAGGGACAGAGGGGACAGAGCA[G/T]GGGAGATGGAGGGAGGGGGTGTGGG
Phenotype
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
GNG8 PubMed Links

Gene Details

Gene
GNG8
Gene Name
G protein subunit gamma 8
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_033258.1 24 Intron NP_150283.1
XM_011527524.2 24 UTR 5 XP_011525826.1
XM_017027503.1 24 Intron XP_016882992.1
XM_017027504.1 24 Intron XP_016882993.1
XM_017027505.1 24 Intron XP_016882994.1

View Full Product Details