Product Details

SNP ID
rs11553205
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:48965536 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGCTCCCAGATTCGTCAGAATTATT[C/T]CACCGACGTGGAGGCAGCCGTCAAC
Phenotype
MIM: 600040 MIM: 134790 MIM: 138570
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
BAX PubMed Links

Gene Details

Gene
BAX
Gene Name
BCL2 associated X, apoptosis regulator
There are no transcripts associated with this gene.

Gene
FTL
Gene Name
ferritin light chain
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000146.3 Intron NP_000137.2
Gene
GYS1
Gene Name
glycogen synthase 1
There are no transcripts associated with this gene.

View Full Product Details