Product Details

SNP ID
rs8105032
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:55270321 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GCCAGCAAAACTCTGCTAGCCCCGG[C/G]GCGAGCCATGGTGGAAGCCCATCTC
Phenotype
MIM: 612939
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
HSPBP1 PubMed Links

Gene Details

Gene
HSPBP1
Gene Name
HSPA (Hsp70) binding protein 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001130106.1 Intron NP_001123578.1
NM_001297600.1 Intron NP_001284529.1
NM_012267.4 Intron NP_036399.3
XM_005258700.2 Intron XP_005258757.1
XM_005258701.2 Intron XP_005258758.1
XM_011526691.1 Intron XP_011524993.1
XM_017026543.1 Intron XP_016882032.1
XM_017026544.1 Intron XP_016882033.1
XM_017026545.1 Intron XP_016882034.1

View Full Product Details