Product Details
- SNP ID
-
rs8110779
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.19:57417227 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- GGAAATAGCGGATTTGGTCAGGTGG[C/T]AGATGCACAGTTGAAGGGAGAGCAT
- Phenotype
-
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
ZNF17
PubMed Links
Gene Details
- Gene
- ZNF17
- Gene Name
- zinc finger protein 17
View Full Product Details