Product Details

SNP ID
rs11559190
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.19:10106572 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCGAGGCCTATGCCGCGAACCCGCA[C/T]TCGTTCGTGTTCACGCGAGGCTGCA
Phenotype
MIM: 609336 MIM: 603913 MIM: 602697 MIM: 607793
Polymorphism
C/T, Transition Substitution
Allele Nomenclature
Literature Links
ANGPTL6 PubMed Links

Gene Details

Gene
ANGPTL6
Gene Name
angiopoietin like 6
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001321411.1 262 Intron NP_001308340.1
NM_031917.2 262 Intron NP_114123.2
XM_005260091.3 262 Intron XP_005260148.1
XM_011528347.2 262 Intron XP_011526649.1
XM_011528348.2 262 Intron XP_011526650.1
XM_011528349.2 262 Intron XP_011526651.1
XM_011528350.2 262 Intron XP_011526652.1
XM_017027347.1 262 Intron XP_016882836.1
Gene
EIF3G
Gene Name
eukaryotic translation initiation factor 3 subunit G
There are no transcripts associated with this gene.

Gene
P2RY11
Gene Name
purinergic receptor P2Y11
There are no transcripts associated with this gene.

Gene
PPAN
Gene Name
peter pan homolog (Drosophila)
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_020230.5 262 Silent Mutation CAC,CAT H30H NP_064615.3
Gene
PPAN-P2RY11
Gene Name
PPAN-P2RY11 readthrough
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001040664.2 262 Silent Mutation CAC,CAT H30H NP_001035754.1
NM_001198690.1 262 Silent Mutation CAC,CAT H30H NP_001185619.1
Gene
SNORD105
Gene Name
small nucleolar RNA, C/D box 105
There are no transcripts associated with this gene.

Gene
SNORD105B
Gene Name
small nucleolar RNA, C/D box 105B
There are no transcripts associated with this gene.

View Full Product Details