Product Details
- SNP ID
-
rs11539839
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.19:17319604 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GAGCTGGAAGGCGAGAGCTCCGCAC[C/T]TCCCCGCAAGGTGGCGCGGACCGAC
- Phenotype
-
MIM: 610216
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
ANO8
PubMed Links
Gene Details
- Gene
- ANO8
- Gene Name
- anoctamin 8
There are no transcripts associated with this gene.
- Gene
- DDA1
- Gene Name
- DET1 and DDB1 associated 1
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_024050.5 |
384 |
Missense Mutation |
CCT,CTT |
P86L |
NP_076955.1 |
View Full Product Details