Product Details

SNP ID
rs11553805
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:17862141 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGGTGAAGAACTTCGGGATCTGGCT[C/G]CGCTATGACTCCCGGAGCGGCACCC
Phenotype
MIM: 604178 MIM: 601843
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
RPL18A PubMed Links

Gene Details

Gene
RPL18A
Gene Name
ribosomal protein L18a
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000980.3 Intron NP_000971.1
Gene
SLC5A5
Gene Name
solute carrier family 5 member 5
There are no transcripts associated with this gene.

Gene
SNORA68
Gene Name
small nucleolar RNA, H/ACA box 68
There are no transcripts associated with this gene.

View Full Product Details