Product Details

SNP ID
rs11554160
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:18173937 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGCAGGCGTCCCCTCTGCAAGCGTT[A/G]GACTTCTTTGGGAATGGGCCACCAG
Phenotype
MIM: 604664 MIM: 603157
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
IFI30 PubMed Links

Gene Details

Gene
IFI30
Gene Name
IFI30, lysosomal thiol reductase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006332.4 158 Silent Mutation TTA,TTG L32L NP_006323.2
Gene
PIK3R2
Gene Name
phosphoinositide-3-kinase regulatory subunit 2
There are no transcripts associated with this gene.

View Full Product Details