Product Details

SNP ID
rs11553483
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:50942085 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATATACACCTCCATTTCCTTTGTAG[A/C]GAGTTCCAGAGACAATATCAAAATT
Phenotype
MIM: 611386 MIM: 603503
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
ADNP PubMed Links

Gene Details

Gene
ADNP
Gene Name
activity dependent neuroprotector homeobox
There are no transcripts associated with this gene.

Gene
ADNP-AS1
Gene Name
ADNP antisense RNA 1
There are no transcripts associated with this gene.

Gene
DPM1
Gene Name
dolichyl-phosphate mannosyltransferase polypeptide 1, catalytic subunit
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001317034.1 481 Missense Mutation NP_001303963.1
NM_001317035.1 481 Missense Mutation NP_001303964.1
NM_001317036.1 481 Missense Mutation NP_001303965.1
NM_003859.2 481 Missense Mutation NP_003850.1
XM_011529093.2 481 Missense Mutation XP_011527395.1

View Full Product Details